Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270934.1 | NP_001257863.1 | 2 - 3 | 664 | 74 |
NM_001270941.1 | NP_001257870.1 | 2 - 3 | 664 | 74 | |
NM_014790.4 | NP_055605.2 | 2 - 3 | 664 | 74 | |
XM_017010097.1 | XP_016865586.1 | 3 - 4 | 658 | 74 | |
XM_017010098.1 | XP_016865587.1 | 2 - 3 | 664 | 74 | |
GenBank mRNA | AB011127.1 | - | 2 - 3 | 358 | 74 |
AF273057.1 | - | 2 - 3 | 505 | 74 | |
AK291280.1 | - | 2 - 3 | 386 | 74 | |
BC003189.1 | - | 2 - 3 | 310 | 74 | |
BC017354.2 | - | 2 - 3 | 429 | 74 | |
EF512550.1 | - | 1 - 2 | 123 | 74 |