Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270934.1 | NP_001257863.1 | 5 - 6 | 1476 | 76 |
NM_001270941.1 | NP_001257870.1 | 5 - 6 | 1476 | 76 | |
NM_001282282.1 | NP_001269211.1 | 4 - 5 | 1199 | 76 | |
NM_014790.4 | NP_055605.2 | 5 - 6 | 1476 | 76 | |
XM_017010097.1 | XP_016865586.1 | 6 - 7 | 1470 | 76 | |
XM_017010098.1 | XP_016865587.1 | 5 - 6 | 1476 | 76 | |
GenBank mRNA | AB011127.1 | - | 5 - 6 | 1170 | 76 |
AF273057.1 | - | 5 - 6 | 1317 | 76 | |
AK291280.1 | - | 5 - 6 | 1198 | 76 | |
AK299724.1 | - | 4 - 5 | 1125 | 76 | |
BC003189.1 | - | 5 - 6 | 1122 | 76 | |
BC017354.2 | - | 5 - 6 | 1241 | 76 | |
EF512550.1 | - | 4 - 5 | 935 | 76 |