Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171038.1 | NP_001164509.1 | 2 - 3 | 399 | 120 |
NM_001171039.1 | NP_001164510.1 | 2 - 3 | 314 | 120 | |
NM_004043.2 | NP_004034.2 | 3 - 4 | 392 | 120 | |
GenBank mRNA | AK314922.1 | - | 2 - 3 | 400 | 120 |
BC001620.1 | - | 2 - 3 | 272 | 120 | |
KJ534774.1 | - | 2 - 3 | 247 | 120 | |
KJ534775.1 | - | 2 - 3 | 247 | 120 | |
KJ534776.1 | - | 2 - 3 | 247 | 120 | |
KJ535045.1 | - | 2 - 3 | 247 | 120 | |
M83779.1 | - | 2 - 3 | 316 | 120 | |
U11090.1 | - | 3 - 4 | 392 | 120 | |
U11091.1 | - | 2 - 3 | 350 | 120 |