Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004661.3 | NP_004652.2 | 8 - 9 | 960 | 93 |
GenBank mRNA | AB011472.1 | - | 8 - 9 | 947 | 93 |
AF053977.1 | - | 8 - 9 | 927 | 93 | |
AF191341.1 | - | 8 - 9 | 911 | 93 | |
AK223250.1 | - | 8 - 9 | 948 | 93 | |
AK291610.1 | - | 8 - 9 | 942 | 93 | |
AK304635.1 | - | 7 - 8 | 824 | 93 | |
BC017713.1 | - | 8 - 9 | 919 | 93 | |
BT009810.1 | - | 8 - 9 | 911 | 93 |