Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130029.1 | NP_001123501.1 | 2 - 3 | 1031 | 63 |
NM_173828.4 | NP_776189.3 | 1 - 2 | 1459 | 63 | |
XM_005268414.3 | XP_005268471.1 | 2 - 3 | 1035 | 63 | |
XM_011537624.1 | XP_011535926.1 | 2 - 3 | 1051 | 63 | |
XM_011537625.2 | XP_011535927.1 | 1 - 2 | 1463 | 63 | |
GenBank mRNA | AK054889.1 | - | 1 - 2 | 1459 | 63 |
AK075064.1 | - | 1 - 2 | 286 | 63 | |
AY358163.1 | - | 1 - 2 | 1395 | 63 | |
BC063469.1 | - | 2 - 3 | 500 | 63 |