Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258426.1 | NP_001245355.1 | 3 - 4 | 542 | 138 |
NM_001258427.1 | NP_001245356.1 | 3 - 4 | 542 | 138 | |
NM_001258428.1 | NP_001245357.1 | 3 - 4 | 614 | 138 | |
NM_031467.2 | NP_113655.2 | 3 - 4 | 542 | 138 | |
XM_005268521.3 | XP_005268578.2 | 3 - 4 | 1099 | 138 | |
XM_011537694.2 | XP_011535996.1 | 3 - 4 | 1171 | 138 | |
XM_017009933.1 | XP_016865422.1 | 3 - 4 | 1171 | 138 | |
XM_017009934.1 | XP_016865423.1 | 3 - 4 | 1171 | 138 | |
XM_017009935.1 | XP_016865424.1 | 3 - 4 | 1171 | 138 | |
XM_017009936.1 | XP_016865425.1 | 3 - 4 | 1099 | 138 | |
XM_017009938.1 | XP_016865427.1 | 3 - 4 | 1171 | 138 | |
XM_017009939.1 | XP_016865428.1 | 3 - 4 | 1099 | 138 | |
XM_017009940.1 | XP_016865429.1 | 3 - 4 | 1171 | 138 | |
XM_017009941.1 | XP_016865430.1 | 3 - 4 | 1171 | 138 | |
XM_017009942.1 | XP_016865431.1 | 3 - 4 | 1171 | 138 | |
GenBank mRNA | AB032762.1 | - | 3 - 4 | 501 | 138 |
AF313465.1 | - | 3 - 4 | 601 | 138 | |
AF332961.1 | - | 3 - 4 | 542 | 138 | |
AF336237.1 | - | 3 - 4 | 534 | 138 | |
BC136262.1 | - | 3 - 4 | 533 | 138 | |
BC143602.1 | - | 3 - 4 | 533 | 138 |