Assay Design



Gene Transcripts

Gene Symbol
SLC4A9
Entrez Gene ID
83697
Gene Name
solute carrier family 4 member 9
Gene Aliases
AE4
Chromosome Location
Chr.5: 140359645 - 140375177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258426.1 NP_001245355.1 8 - 9 1114 68
XM_017009935.1 XP_016865424.1 8 - 9 1743 68
XM_017009939.1 XP_016865428.1 8 - 9 1671 68
GenBank mRNA AF336237.1 - 8 - 9 1106 68

View Full Product Details