Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018037.1 | NP_001018047.1 | 55 - 56 | 8098 | 148 |
NM_001018038.2 | NP_001018048.1 | 56 - 57 | 8215 | 148 | |
NM_015186.3 | NP_056001.1 | 56 - 57 | 8215 | 148 | |
NM_033305.2 | NP_150648.2 | 56 - 57 | 8215 | 148 | |
GenBank mRNA | AB023203.1 | - | 17 - 18 | 3046 | 148 |
AB054005.1 | - | 56 - 57 | 8126 | 148 | |
AF337532.1 | - | 56 - 57 | 8215 | 148 | |
AJ608769.1 | - | 55 - 56 | 8098 | 148 | |
AJ626859.1 | - | 56 - 57 | 8215 | 148 | |
AK022967.1 | - | 9 - 10 | 1373 | 148 | |
AK027023.1 | - | 3 - 4 | 517 | 148 | |
BC041852.1 | - | 14 - 15 | 2410 | 148 |