Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018037.1 | NP_001018047.1 | 7 - 8 | 813 | 140 |
NM_001018038.2 | NP_001018048.1 | 7 - 8 | 813 | 140 | |
NM_015186.3 | NP_056001.1 | 7 - 8 | 813 | 140 | |
NM_033305.2 | NP_150648.2 | 7 - 8 | 813 | 140 | |
GenBank mRNA | AB054005.1 | - | 7 - 8 | 724 | 140 |
AF337532.1 | - | 7 - 8 | 813 | 140 | |
AJ608769.1 | - | 7 - 8 | 813 | 140 | |
AJ626859.1 | - | 7 - 8 | 813 | 140 |