Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260502.1 | NP_001247431.1 | 8 - 9 | 992 | 81 |
NM_001260503.1 | NP_001247432.1 | 2 - 3 | 301 | 81 | |
NM_014230.3 | NP_055045.2 | 9 - 10 | 1106 | 81 | |
NR_048541.1 | 8 - 9 | 1002 | 81 | ||
GenBank mRNA | AF195951.1 | - | 10 - 11 | 1047 | 81 |
AK027320.1 | - | 4 - 5 | 342 | 81 | |
AK027322.1 | - | - | 274 | 81 | |
AK074698.1 | - | 9 - 10 | 1082 | 81 | |
AK097962.1 | - | 2 - 3 | 301 | 81 | |
AK126258.1 | - | 8 - 9 | 973 | 81 | |
AK301100.1 | - | 8 - 9 | 973 | 81 | |
AL512708.1 | - | 9 - 10 | 1021 | 81 | |
BC020238.1 | - | 8 - 9 | 983 | 81 |