Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260502.1 | NP_001247431.1 | 14 - 15 | 1581 | 65 |
NM_001260503.1 | NP_001247432.1 | 8 - 9 | 890 | 65 | |
NM_014230.3 | NP_055045.2 | 15 - 16 | 1695 | 65 | |
NR_048541.1 | 14 - 15 | 1591 | 65 | ||
GenBank mRNA | AF195951.1 | - | 16 - 17 | 1636 | 65 |
AK002002.1 | - | 10 - 11 | 1150 | 65 | |
AK027322.1 | - | - | 863 | 65 | |
AK074698.1 | - | 15 - 16 | 1671 | 65 | |
AK092935.1 | - | 1 - 2 | 1214 | 65 | |
AK097962.1 | - | 8 - 9 | 890 | 65 | |
AK126258.1 | - | 14 - 15 | 1562 | 65 | |
AK301100.1 | - | 14 - 15 | 1562 | 65 | |
AL512708.1 | - | 15 - 16 | 1610 | 65 | |
BC020238.1 | - | 14 - 15 | 1572 | 65 | |
DQ786315.1 | - | 3 - 4 | 266 | 65 |