Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014358.3 | NP_055173.1 | 2 - 3 | 300 | 102 |
XM_011520615.1 | XP_011518917.1 | 2 - 3 | 239 | 102 | |
GenBank mRNA | AB024718.1 | - | 2 - 3 | 286 | 102 |
AK312671.1 | - | 2 - 3 | 301 | 102 | |
AY358499.1 | - | 2 - 3 | 240 | 102 | |
BC000715.2 | - | 2 - 3 | 218 | 102 | |
DA678013.1 | - | 2 - 3 | 301 | 102 |