Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014358.3 | NP_055173.1 | 4 - 5 | 536 | 97 |
XM_011520614.2 | XP_011518916.1 | 3 - 4 | 809 | 97 | |
GenBank mRNA | AB024718.1 | - | 4 - 5 | 522 | 97 |
AK312671.1 | - | 4 - 5 | 537 | 97 | |
AY358499.1 | - | 4 - 5 | 476 | 97 | |
BC000715.2 | - | 4 - 5 | 454 | 97 |