Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270697.1 | NP_001257626.1 | 9 - 10 | 1610 | 104 |
NM_018287.6 | NP_060757.4 | 10 - 11 | 1751 | 104 | |
XM_011519761.1 | XP_011518063.1 | 8 - 9 | 1529 | 104 | |
GenBank mRNA | AK127611.1 | - | 9 - 10 | 1652 | 104 |
AY033595.1 | - | 10 - 11 | 1718 | 104 | |
BC115362.2 | - | 9 - 10 | 1507 | 104 | |
BC115363.2 | - | 9 - 10 | 1507 | 104 |