Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270697.1 | NP_001257626.1 | 8 - 9 | 1546 | 124 |
NM_018287.6 | NP_060757.4 | 9 - 10 | 1687 | 124 | |
XM_011519761.1 | XP_011518063.1 | 7 - 8 | 1465 | 124 | |
GenBank mRNA | AK127611.1 | - | 8 - 9 | 1588 | 124 |
AY033595.1 | - | 9 - 10 | 1654 | 124 | |
BC115362.2 | - | 8 - 9 | 1443 | 124 | |
BC115363.2 | - | 8 - 9 | 1443 | 124 |