Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258452.1 | NP_001245381.1 | 7 - 8 | 1478 | 81 |
NM_001258453.1 | NP_001245382.1 | 8 - 9 | 1550 | 81 | |
NM_032807.4 | NP_116196.3 | 8 - 9 | 1566 | 81 | |
NM_178150.2 | NP_835363.1 | 7 - 8 | 1452 | 81 | |
XM_011519748.2 | XP_011518050.1 | 9 - 10 | 2146 | 81 | |
XM_017016842.1 | XP_016872331.1 | 8 - 9 | 2693 | 81 | |
XM_017016843.1 | XP_016872332.1 | 9 - 10 | 2077 | 81 | |
XM_017016844.1 | XP_016872333.1 | 7 - 8 | 1462 | 81 | |
XM_017016845.1 | XP_016872334.1 | 7 - 8 | 1819 | 81 | |
XM_017016846.1 | XP_016872335.1 | 6 - 7 | 1271 | 81 | |
GenBank mRNA | AF380349.1 | - | 8 - 9 | 1566 | 81 |
AF454502.1 | - | 7 - 8 | 1393 | 81 | |
AK027496.1 | - | 4 - 5 | 535 | 81 | |
AK095343.1 | - | 7 - 8 | 1426 | 81 | |
AK122753.1 | - | 7 - 8 | 1478 | 81 | |
AK226128.1 | - | 7 - 8 | 1976 | 81 | |
AK315173.1 | - | 6 - 7 | 1168 | 81 | |
BC020266.1 | - | 5 - 6 | 960 | 81 | |
BC032674.1 | - | 5 - 6 | 904 | 81 | |
BC080629.1 | - | 5 - 6 | 1115 | 81 | |
BC110884.1 | - | 8 - 9 | 1475 | 81 | |
BC113377.1 | - | 7 - 8 | 1399 | 81 |