Assay Design
Gene Transcripts
- Gene Symbol
- BSCL2
- Entrez Gene ID
-
26580
- Gene Name
- BSCL2, seipin lipid droplet biogenesis associated
- Gene Aliases
-
GNG3LG, HMN5, PELD, SPG17
- Chromosome Location
-
Chr.11: 62690262 - 62709619 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037949.1 |
|
9 - 10 |
1673 |
64 |
GenBank mRNA |
BC009866.2 |
- |
9 - 10 |
1568 |
64 |
View Full Product Details