Assay Design



Gene Transcripts

Gene Symbol
KNG1
Entrez Gene ID
3827
Gene Name
kininogen 1
Gene Aliases
BDK, BK, KNG
Chromosome Location
Chr.3: 186717309 - 186744410 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000893.3 NP_000884.1 7 - 8 1169 121
NM_001102416.2 NP_001095886.1 7 - 8 1169 121
NM_001166451.1 NP_001159923.1 6 - 7 1061 121
GenBank mRNA AK223589.1 - 7 - 8 1118 121
AK298440.1 - 8 - 9 1083 121
AK303768.1 - 7 - 8 1149 121
AK315230.1 - 7 - 8 1121 121
BC026253.1 - 7 - 8 847 121
BC060039.1 - 7 - 8 1072 121
CD014130.1 - 6 - 7 1023 121
K02566.1 - 7 - 8 984 121

View Full Product Details