Assay Design



Gene Transcripts

Gene Symbol
HPS6
Entrez Gene ID
79803
Gene Name
HPS6, biogenesis of lysosomal organelles complex 2 subunit 3
Gene Aliases
BLOC2S3
Chromosome Location
Chr.10: 102065367 - 102068038 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024747.5 NP_079023.2 1 - 1 849 129
GenBank mRNA AF536238.1 - 1 - 1 814 129
BC011594.1 - 1 - 1 814 129
BC014993.1 - 1 - 1 826 129

View Full Product Details