Assay Design



Gene Transcripts

Gene Symbol
PRRC2C
Entrez Gene ID
23215
Gene Name
proline rich coiled-coil 2C
Gene Aliases
BAT2-iso, BAT2D1, BAT2L2, XTP2
Chromosome Location
Chr.1: 171485496 - 171593511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015172.3 NP_055987.2 14 - 15 2554 111
XM_005245015.2 XP_005245072.1 14 - 15 2591 111
XM_005245016.4 XP_005245073.1 14 - 15 2571 111
XM_005245017.2 XP_005245074.1 14 - 15 2591 111
XM_005245018.2 XP_005245075.1 14 - 15 2566 111
XM_005245019.2 XP_005245076.1 14 - 15 2591 111
XM_005245020.2 XP_005245077.1 14 - 15 2591 111
XM_005245023.2 XP_005245080.1 14 - 15 2591 111
XM_005245024.2 XP_005245081.1 14 - 15 2591 111
XM_005245025.2 XP_005245082.1 14 - 15 2591 111
XM_005245026.2 XP_005245083.1 14 - 15 2591 111
XM_005245027.2 XP_005245084.1 14 - 15 2591 111
XM_005245028.2 XP_005245085.1 14 - 15 2591 111
XM_005245029.2 XP_005245086.1 14 - 15 2591 111
XM_006711238.3 XP_006711301.1 14 - 15 2571 111
XM_017000760.1 XP_016856249.1 14 - 15 2556 111
XM_017000761.1 XP_016856250.1 14 - 15 2591 111
XM_017000762.1 XP_016856251.1 14 - 15 2591 111
GenBank mRNA AF488829.1 - 13 - 14 2288 111
AK297263.1 - 3 - 4 589 111
AK298887.1 - - 1585 111
AL096857.1 - 13 - 14 2345 111

View Full Product Details