Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133494.2 | NP_598001.1 | 7 - 8 | 917 | 129 |
XM_017000344.1 | XP_016855833.1 | 7 - 8 | 919 | 129 | |
XM_017000345.1 | XP_016855834.1 | 8 - 9 | 1026 | 129 | |
XM_017000346.1 | XP_016855835.1 | 7 - 8 | 919 | 129 | |
GenBank mRNA | AB062450.1 | - | 6 - 7 | 636 | 129 |
AK313411.1 | - | 7 - 8 | 717 | 129 |