Assay Design



Gene Transcripts

Gene Symbol
NEK7
Entrez Gene ID
140609
Gene Name
NIMA related kinase 7
Gene Aliases
-
Chromosome Location
Chr.1: 198156978 - 198322420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017000345.1 XP_016855834.1 2 - 3 385 136

View Full Product Details