Assay Design



Gene Transcripts

Gene Symbol
LPP
Entrez Gene ID
4026
Gene Name
LIM domain containing preferred translocation partner in lipoma
Gene Aliases
-
Chromosome Location
Chr.3: 188153050 - 188890671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006381.1 XP_016861870.1 7 - 8 1429 134

View Full Product Details