Assay Design



Gene Transcripts

Gene Symbol
ARL5B
Entrez Gene ID
221079
Gene Name
ADP ribosylation factor like GTPase 5B
Gene Aliases
ARL8
Chromosome Location
Chr.10: 18659335 - 18681639 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178815.3 NP_848930.1 1 - 2 301 76
XM_005252400.1 XP_005252457.1 1 - 2 350 76
GenBank mRNA AF494061.1 - 1 - 2 223 76
AK027275.1 - 1 - 2 254 76
BC024163.1 - 1 - 2 281 76

View Full Product Details