Assay Design



Gene Transcripts

Gene Symbol
NEK7
Entrez Gene ID
140609
Gene Name
NIMA related kinase 7
Gene Aliases
-
Chromosome Location
Chr.1: 198156978 - 198322420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017000347.1 XP_016855836.1 7 - 8 912 84
XM_017000348.1 XP_016855837.1 7 - 8 912 84

View Full Product Details