Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008844.2 | NP_001008844.1 | 22 - 23 | 3422 | 133 |
NM_001319034.1 | NP_001305963.1 | 22 - 23 | 3422 | 133 | |
NM_004415.3 | NP_004406.2 | 22 - 23 | 3422 | 133 | |
GenBank mRNA | AB209992.1 | - | 22 - 23 | 3380 | 133 |
AK296756.1 | - | 20 - 21 | 2624 | 133 | |
AK304182.1 | - | 22 - 23 | 3325 | 133 | |
BC140802.1 | - | 22 - 23 | 3355 | 133 | |
HM151899.1 | - | 22 - 23 | 3360 | 133 | |
M77830.3 | - | 22 - 23 | 3360 | 133 |