Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008844.2 | NP_001008844.1 | 9 - 10 | 1473 | 87 |
NM_001319034.1 | NP_001305963.1 | 9 - 10 | 1473 | 87 | |
NM_004415.3 | NP_004406.2 | 9 - 10 | 1473 | 87 | |
GenBank mRNA | AB209992.1 | - | 9 - 10 | 1431 | 87 |
AK296756.1 | - | 7 - 8 | 675 | 87 | |
AK301676.1 | - | 7 - 8 | 656 | 87 | |
AK304182.1 | - | 9 - 10 | 1376 | 87 | |
BC140802.1 | - | 9 - 10 | 1406 | 87 | |
HM151899.1 | - | 9 - 10 | 1411 | 87 | |
M77830.3 | - | 9 - 10 | 1411 | 87 |