Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001657.3 | NP_001648.1 | 2 - 3 | 560 | 113 |
GenBank mRNA | BC009799.1 | - | 2 - 3 | 526 | 113 |
BC146953.1 | - | 2 - 3 | 367 | 113 | |
BC146967.1 | - | 2 - 3 | 367 | 113 | |
BT019866.1 | - | 2 - 3 | 312 | 113 | |
M30704.1 | - | 2 - 3 | 521 | 113 |