Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319042.1 | NP_001305971.1 | 2 - 3 | 309 | 156 |
NM_001446.4 | NP_001437.1 | 2 - 3 | 568 | 156 | |
GenBank mRNA | AJ002962.1 | - | 2 - 3 | 324 | 156 |
AK311867.1 | - | 2 - 3 | 327 | 156 | |
AW156936.1 | - | 2 - 3 | 156 | ||
BC012299.1 | - | 2 - 3 | 434 | 156 | |
BU736791.1 | - | 2 - 3 | 156 | ||
D50373.1 | - | 2 - 3 | 300 | 156 | |
D88648.1 | - | 2 - 3 | 299 | 156 | |
KM111186.1 | - | 2 - 3 | 309 | 156 | |
U51338.1 | - | 2 - 3 | 303 | 156 | |
U81235.1 | - | 2 - 3 | 263 | 156 |