Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020728.2 | NP_065779.1 | 21 - 22 | 2700 | 67 |
GenBank mRNA | AB033054.2 | - | 22 - 23 | 2486 | 67 |
AK001181.1 | - | 10 - 11 | 1189 | 67 | |
AK026710.1 | - | 6 - 7 | 1120 | 67 | |
AK124091.1 | - | 6 - 7 | 1446 | 67 | |
AK126214.1 | - | 22 - 23 | 2221 | 67 | |
AK308824.1 | - | 21 - 22 | 2308 | 67 | |
AL833233.1 | - | 21 - 22 | 2309 | 67 | |
AY368150.1 | - | 21 - 22 | 2700 | 67 | |
BC013957.2 | - | 13 - 14 | 1422 | 67 | |
DQ993201.1 | - | 23 - 24 | 2718 | 67 |