Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290184.1 | NP_001277113.1 | 13 - 14 | 1716 | 64 |
NM_001290186.1 | NP_001277115.1 | 9 - 10 | 1163 | 64 | |
NM_015331.2 | NP_056146.1 | 12 - 13 | 1577 | 64 | |
XM_011509363.2 | XP_011507665.1 | 12 - 13 | 1794 | 64 | |
GenBank mRNA | AF240468.1 | - | 12 - 13 | 1595 | 64 |
AK296153.1 | - | 7 - 8 | 862 | 64 | |
AK299142.1 | - | 9 - 10 | 1050 | 64 | |
AK314764.1 | - | 12 - 13 | 1484 | 64 | |
AY359120.1 | - | 12 - 13 | 1454 | 64 | |
BC047621.1 | - | 13 - 14 | 1673 | 64 | |
D87442.1 | - | 12 - 13 | 1451 | 64 |