Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290184.1 | NP_001277113.1 | 9 - 10 | 1259 | 62 |
NM_015331.2 | NP_056146.1 | 8 - 9 | 1120 | 62 | |
XM_005245053.4 | XP_005245110.1 | 8 - 9 | 1337 | 62 | |
XM_011509363.2 | XP_011507665.1 | 8 - 9 | 1337 | 62 | |
GenBank mRNA | AF240468.1 | - | 8 - 9 | 1138 | 62 |
AK296153.1 | - | 3 - 4 | 405 | 62 | |
AK314764.1 | - | 8 - 9 | 1027 | 62 | |
AY359120.1 | - | 8 - 9 | 997 | 62 | |
BC047621.1 | - | 9 - 10 | 1216 | 62 | |
D87442.1 | - | 8 - 9 | 994 | 62 |