Assay Design
Gene Transcripts
- Gene Symbol
- COL10A1
- Entrez Gene ID
-
1300
- Gene Name
- collagen type X alpha 1 chain
- Gene Aliases
-
-
- Chromosome Location
-
Chr.6: 116118909 - 116218698 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_000493.3 |
NP_000484.2 |
1 - 2 |
82 |
66 |
View Full Product Details