Assay Design



Gene Transcripts

Gene Symbol
CUL3
Entrez Gene ID
8452
Gene Name
cullin 3
Gene Aliases
CUL-3, PHA2E
Chromosome Location
Chr.2: 224470150 - 224585397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257197.1 NP_001244126.1 11 - 12 1903 78
NM_001257198.1 NP_001244127.1 12 - 13 1784 78
NM_003590.4 NP_003581.1 12 - 13 2101 78
XM_006712800.2 XP_006712863.2 12 - 13 1774 78
XM_011511994.2 XP_011510296.1 11 - 12 1658 78
XM_011511995.1 XP_011510297.1 12 - 13 1686 78
XM_011511996.2 XP_011510298.1 12 - 13 1750 78
GenBank mRNA AB014517.2 - 12 - 13 2029 78
AF052147.1 - 5 - 6 524 78
AF062537.1 - 12 - 13 1802 78
AF064087.1 - 12 - 13 1794 78
AK291151.1 - 12 - 13 1751 78
AK299644.1 - 11 - 12 1513 78
AK308496.1 - 12 - 13 1784 78
AY337761.1 - 12 - 13 2247 78
BC031844.1 - 5 - 6 529 78
BC039598.1 - 12 - 13 2046 78
BC092409.1 - 12 - 13 2046 78
U58089.1 - 8 - 9 1140 78

View Full Product Details