Assay Design



Gene Transcripts

Gene Symbol
CUL3
Entrez Gene ID
8452
Gene Name
cullin 3
Gene Aliases
CUL-3, PHA2E
Chromosome Location
Chr.2: 224470150 - 224585397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257197.1 NP_001244126.1 7 - 8 1400 157
NM_001257198.1 NP_001244127.1 8 - 9 1281 157
NM_003590.4 NP_003581.1 8 - 9 1598 157
XM_006712800.2 XP_006712863.2 8 - 9 1271 157
XM_011511994.2 XP_011510296.1 7 - 8 1155 157
XM_011511995.1 XP_011510297.1 8 - 9 1183 157
XM_011511996.2 XP_011510298.1 8 - 9 1247 157
GenBank mRNA AB014517.2 - 8 - 9 1526 157
AF062537.1 - 8 - 9 1299 157
AF064087.1 - 8 - 9 1291 157
AK291151.1 - 8 - 9 1248 157
AK299644.1 - 7 - 8 1010 157
AK308496.1 - 8 - 9 1281 157
AY337761.1 - 8 - 9 1744 157
BC039598.1 - 8 - 9 1543 157
BC092409.1 - 8 - 9 1543 157
U58089.1 - 4 - 5 637 157

View Full Product Details