Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034852.2 | NP_001030024.1 | 3 - 4 | 661 | 67 |
NM_022137.5 | NP_071420.1 | 3 - 4 | 661 | 67 | |
XM_005267995.1 | XP_005268052.1 | 3 - 4 | 670 | 67 | |
XM_005267996.1 | XP_005268053.1 | 3 - 4 | 670 | 67 | |
GenBank mRNA | AJ249900.1 | - | 3 - 4 | 633 | 67 |
AK289988.1 | - | 3 - 4 | 642 | 67 | |
AK313063.1 | - | 3 - 4 | 642 | 67 | |
BC008608.1 | - | 3 - 4 | 629 | 67 | |
BC011548.1 | - | 3 - 4 | 632 | 67 |