Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161498.1 | NP_001154970.1 | 10 - 11 | 1264 | 76 |
XM_011536762.2 | XP_011535064.1 | 7 - 8 | 710 | 76 | |
XM_011536763.2 | XP_011535065.1 | 5 - 6 | 570 | 76 | |
XM_017021290.1 | XP_016876779.1 | 10 - 11 | 1626 | 76 | |
GenBank mRNA | BC146872.1 | - | 10 - 11 | 1169 | 76 |