Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 4 - 5 | 757 | 67 |
XM_005270276.4 | XP_005270333.1 | 4 - 5 | 711 | 67 | |
XM_006718056.3 | XP_006718119.1 | 4 - 5 | 720 | 67 | |
XM_006718057.3 | XP_006718120.1 | 4 - 5 | 727 | 67 | |
XM_011540341.2 | XP_011538643.1 | 4 - 5 | 881 | 67 | |
XM_011540342.1 | XP_011538644.1 | 4 - 5 | 459 | 67 | |
XM_011540344.2 | XP_011538646.1 | 3 - 4 | 366 | 67 | |
XM_011540345.1 | XP_011538647.1 | 3 - 4 | 491 | 67 | |
XM_011540346.2 | XP_011538648.1 | 4 - 5 | 726 | 67 | |
XM_017016913.1 | XP_016872402.1 | 4 - 5 | 420 | 67 | |
XM_017016914.1 | XP_016872403.1 | 4 - 5 | 750 | 67 | |
GenBank mRNA | AB011155.2 | - | 4 - 5 | 787 | 67 |
AB091676.1 | - | 2 - 3 | 222 | 67 | |
AK122931.1 | - | 4 - 5 | 462 | 67 | |
BC146794.1 | - | 4 - 5 | 787 | 67 |