Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021095.2 | NP_066918.2 | 13 - 14 | 1861 | 86 |
NR_028323.1 | 12 - 13 | 2213 | 86 | ||
XM_006712128.1 | XP_006712191.1 | 13 - 14 | 1656 | 86 | |
XM_006712129.1 | XP_006712192.1 | 13 - 14 | 1704 | 86 | |
XM_006712130.1 | XP_006712193.1 | 13 - 14 | 1695 | 86 | |
XM_011533146.2 | XP_011531448.1 | 11 - 12 | 1421 | 86 | |
XM_017005216.1 | XP_016860705.1 | 10 - 11 | 1431 | 86 | |
GenBank mRNA | AF069307.1 | - | 13 - 14 | 1757 | 86 |
AF081571.1 | - | 12 - 13 | 1573 | 86 | |
AK027855.1 | - | 12 - 13 | 2053 | 86 | |
AK075078.1 | - | 4 - 5 | 299 | 86 | |
AK293162.1 | - | 7 - 8 | 745 | 86 | |
AK314545.1 | - | 14 - 15 | 1688 | 86 | |
BC012806.2 | - | 11 - 12 | 1423 | 86 | |
BC015631.2 | - | 12 - 13 | 1574 | 86 | |
BQ651664.1 | - | 1 - 2 | 100 | 86 | |
DA958474.1 | - | 2 - 3 | 395 | 86 |