Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021095.2 | NP_066918.2 | 14 - 15 | 2037 | 101 |
NR_028323.1 | 13 - 14 | 2389 | 101 | ||
XM_006712128.1 | XP_006712191.1 | 14 - 15 | 1832 | 101 | |
XM_006712129.1 | XP_006712192.1 | 14 - 15 | 1880 | 101 | |
XM_006712130.1 | XP_006712193.1 | 14 - 15 | 1871 | 101 | |
XM_011533146.2 | XP_011531448.1 | 12 - 13 | 1597 | 101 | |
XM_017005216.1 | XP_016860705.1 | 11 - 12 | 1607 | 101 | |
GenBank mRNA | AF069307.1 | - | 14 - 15 | 1933 | 101 |
AF081571.1 | - | 13 - 14 | 1749 | 101 | |
AK075078.1 | - | 5 - 6 | 475 | 101 | |
AK293162.1 | - | 8 - 9 | 921 | 101 | |
AK314545.1 | - | 15 - 16 | 1864 | 101 | |
BC012806.2 | - | 12 - 13 | 1599 | 101 | |
BC015631.2 | - | 13 - 14 | 1750 | 101 | |
BQ651664.1 | - | 2 - 3 | 276 | 101 |