Assay Design



Gene Transcripts

Gene Symbol
PRKCZ
Entrez Gene ID
5590
Gene Name
protein kinase C zeta
Gene Aliases
PKC-ZETA, PKC2
Chromosome Location
Chr.1: 2050470 - 2185395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033581.1 NP_001028753.1 11 - 12 1354 60
NM_001033582.1 NP_001028754.1 11 - 12 1251 60
NM_001242874.1 NP_001229803.1 11 - 12 1361 60
NM_002744.4 NP_002735.3 14 - 15 1566 60
XM_011541773.1 XP_011540075.1 14 - 15 1562 60
XM_011541774.1 XP_011540076.1 11 - 12 1427 60
XM_011541776.1 XP_011540078.1 11 - 12 1367 60
XM_011541778.1 XP_011540080.1 11 - 12 1280 60
XM_017001789.1 XP_016857278.1 14 - 15 1586 60
XM_017001790.1 XP_016857279.1 14 - 15 1586 60
XM_017001791.1 XP_016857280.1 11 - 12 1211 60
XM_017001792.1 XP_016857281.1 11 - 12 1391 60
XM_017001793.1 XP_016857282.1 11 - 12 1649 60
XM_017001794.1 XP_016857283.1 11 - 12 1304 60
XM_017001795.1 XP_016857284.1 11 - 12 1308 60
XM_017001796.1 XP_016857285.1 11 - 12 1391 60
XM_017001797.1 XP_016857286.1 9 - 10 1032 60
XM_017001798.1 XP_016857287.1 11 - 12 1304 60
XM_017001799.1 XP_016857288.1 11 - 12 1688 60
XM_017001800.1 XP_016857289.1 9 - 10 1008 60
XM_017001801.1 XP_016857290.1 9 - 10 1008 60
XM_017001802.1 XP_016857291.1 11 - 12 1564 60
XM_017001804.1 XP_016857293.1 11 - 12 1474 60
XM_017001805.1 XP_016857294.1 11 - 12 1444 60
GenBank mRNA AK097627.1 - 9 - 10 987 60
AK131526.1 - 4 - 5 1101 60
AK290995.1 - 14 - 15 1405 60
AK294649.1 - 11 - 12 1356 60
AK294782.1 - 11 - 12 1274 60
AK302160.1 - 9 - 10 978 60
BC008058.2 - 14 - 15 1513 60
BC014270.2 - 14 - 15 1566 60
BT007082.1 - 14 - 15 1405 60
L14283.1 - 14 - 15 1405 60
Z15108.1 - 14 - 15 1387 60

View Full Product Details