Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002744.4 | NP_002735.3 | 1 - 2 | 229 | 56 |
XM_011541773.1 | XP_011540075.1 | 1 - 2 | 225 | 56 | |
XM_011541775.2 | XP_011540077.1 | 1 - 2 | 225 | 56 | |
XM_017001789.1 | XP_016857278.1 | 1 - 2 | 225 | 56 | |
XM_017001790.1 | XP_016857279.1 | 1 - 2 | 225 | 56 | |
XM_017001803.1 | XP_016857292.1 | 1 - 2 | 225 | 56 | |
GenBank mRNA | AK290995.1 | - | 1 - 2 | 68 | 56 |
BC008058.2 | - | 1 - 2 | 176 | 56 | |
BC014270.2 | - | 1 - 2 | 229 | 56 | |
BT007082.1 | - | 1 - 2 | 68 | 56 | |
L14283.1 | - | 1 - 2 | 68 | 56 | |
Z15108.1 | - | 1 - 2 | 50 | 56 |