Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004949.4 | NP_004940.1 | 9 - 10 | 1726 | 112 |
NM_024422.4 | NP_077740.1 | 9 - 10 | 1726 | 112 | |
XM_005258206.4 | XP_005258263.1 | 9 - 10 | 1227 | 112 | |
GenBank mRNA | AK290313.1 | - | 9 - 10 | 1417 | 112 |
AK297029.1 | - | 10 - 11 | 1386 | 112 | |
BC063291.1 | - | 9 - 10 | 1708 | 112 | |
BX648425.1 | - | 9 - 10 | 1278 | 112 | |
CR749226.1 | - | 9 - 10 | 1445 | 112 | |
EF017811.1 | - | 9 - 10 | 1265 | 112 | |
EF017812.1 | - | 9 - 10 | 1265 | 112 | |
X56807.1 | - | 9 - 10 | 1305 | 112 |