Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001982.3 | NP_001973.2 | 23 - 24 | 3111 | 71 |
GenBank mRNA | AK124710.1 | - | 6 - 7 | 1293 | 71 |
AK125028.1 | - | 1 - 2 | 653 | 71 | |
AK291681.1 | - | 23 - 24 | 2964 | 71 | |
AK294719.1 | - | 22 - 23 | 3151 | 71 | |
AK295650.1 | - | 24 - 25 | 2878 | 71 | |
AK300909.1 | - | 10 - 11 | 1210 | 71 | |
BC041579.1 | - | 8 - 9 | 756 | 71 | |
BC082992.1 | - | 23 - 24 | 2950 | 71 | |
M29366.1 | - | 23 - 24 | 2934 | 71 | |
M34309.1 | - | 23 - 24 | 3033 | 71 |