Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174858.2 | NP_777283.1 | 1 - 2 | 397 | 64 |
XM_005270739.4 | XP_005270796.1 | 1 - 2 | 316 | 64 | |
XM_017001012.1 | XP_016856501.1 | 1 - 2 | 466 | 64 | |
GenBank mRNA | AF445193.1 | - | 1 - 2 | 397 | 64 |
AK090967.1 | - | 1 - 2 | 316 | 64 | |
BC036666.1 | - | 1 - 2 | 323 | 64 |