Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012093.3 | NP_036225.2 | 2 - 3 | 1195 | 65 |
NM_174858.2 | NP_777283.1 | 2 - 3 | 584 | 65 | |
XM_006710572.3 | XP_006710635.1 | 2 - 3 | 362 | 65 | |
XM_017001008.1 | XP_016856497.1 | 2 - 3 | 379 | 65 | |
XM_017001009.1 | XP_016856498.1 | 2 - 3 | 1087 | 65 | |
XM_017001012.1 | XP_016856501.1 | 2 - 3 | 653 | 65 | |
GenBank mRNA | AF445193.1 | - | 2 - 3 | 584 | 65 |
AY171600.2 | - | 2 - 3 | 1200 | 65 | |
BC033896.1 | - | 2 - 3 | 1200 | 65 |