Assay Design



Gene Transcripts

Gene Symbol
NTM
Entrez Gene ID
50863
Gene Name
neurotrimin
Gene Aliases
HNT, IGLON2, NTRI
Chromosome Location
Chr.11: 131370326 - 132336822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048209.1 NP_001041674.1 5 - 6 990 72
NM_001144058.1 NP_001137530.1 4 - 5 1323 72
NM_001144059.1 NP_001137531.1 4 - 5 1323 72
NM_016522.2 NP_057606.1 4 - 5 1323 72
XM_005271580.3 XP_005271637.1 5 - 6 1140 72
XM_005271581.3 XP_005271638.1 4 - 5 1093 72
XM_005271582.3 XP_005271639.1 5 - 6 1140 72
XM_006718855.3 XP_006718918.1 6 - 7 1247 72
XM_006718857.3 XP_006718920.1 5 - 6 1167 72
XM_011542861.2 XP_011541163.1 4 - 5 1093 72
XM_017017851.1 XP_016873340.1 4 - 5 1093 72
XM_017017852.1 XP_016873341.1 5 - 6 1140 72
XM_017017853.1 XP_016873342.1 6 - 7 1247 72
XM_017017854.1 XP_016873343.1 4 - 5 1093 72
XM_017017855.1 XP_016873344.1 5 - 6 1140 72
XM_017017856.1 XP_016873345.1 6 - 7 853 72
XM_017017857.1 XP_016873346.1 6 - 7 1144 72
XM_017017858.1 XP_016873347.1 6 - 7 1137 72
XM_017017859.1 XP_016873348.1 6 - 7 853 72
XM_017017860.1 XP_016873349.1 5 - 6 1170 72
XM_017017861.1 XP_016873350.1 6 - 7 1247 72
XM_017017862.1 XP_016873351.1 6 - 7 1134 72
XM_017017863.1 XP_016873352.1 5 - 6 1170 72
XM_017017864.1 XP_016873353.1 5 - 6 1138 72
XM_017017865.1 XP_016873354.1 5 - 6 1140 72
XM_017017866.1 XP_016873355.1 5 - 6 1167 72
XM_017017867.1 XP_016873356.1 3 - 4 347 72
GenBank mRNA AF126426.1 - 4 - 5 923 72
AK293448.1 - 5 - 6 958 72
AK293487.1 - 6 - 7 939 72
AK294137.1 - 5 - 6 869 72
AK315964.1 - 5 - 6 985 72
AY358331.1 - 5 - 6 792 72
BC050716.1 - 4 - 5 673 72
BE263639.1 - 3 - 4 389 72
EF043567.1 - 4 - 5 679 72

View Full Product Details