Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130860.3 | NP_001124332.1 | 17 - 18 | 3153 | 69 |
NM_001282141.1 | NP_001269070.1 | 14 - 15 | 2577 | 69 | |
NM_014799.3 | NP_055614.1 | 16 - 17 | 2973 | 69 | |
NM_138737.4 | NP_620074.3 | 17 - 18 | 3240 | 69 | |
XM_006724722.1 | XP_006724785.1 | 17 - 18 | 3085 | 69 | |
XM_011531073.2 | XP_011529375.1 | 17 - 18 | 3262 | 69 | |
XM_011531074.2 | XP_011529376.1 | 16 - 17 | 3049 | 69 | |
XM_017029999.1 | XP_016885488.1 | 16 - 17 | 2797 | 69 | |
XM_017030000.1 | XP_016885489.1 | 16 - 17 | 2797 | 69 | |
GenBank mRNA | AB014598.2 | - | 16 - 17 | 2936 | 69 |
AF148860.1 | - | 16 - 17 | 2924 | 69 | |
AJ296162.2 | - | 16 - 17 | 3563 | 69 | |
AK294278.1 | - | 8 - 9 | 1241 | 69 | |
AK295456.1 | - | 8 - 9 | 1238 | 69 | |
AK307761.1 | - | 14 - 15 | 2442 | 69 | |
AK316231.1 | - | 8 - 9 | 1238 | 69 | |
AY358990.1 | - | 17 - 18 | 3035 | 69 | |
BC011561.1 | - | 17 - 18 | 2968 | 69 |