Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024953.3 | NP_079229.2 | 7 - 8 | 717 | 63 |
XM_005253938.1 | XP_005253995.1 | 6 - 7 | 563 | 63 | |
XM_006719606.2 | XP_006719669.1 | 7 - 8 | 1377 | 63 | |
XM_017019977.1 | XP_016875466.1 | 5 - 6 | 440 | 63 | |
GenBank mRNA | AB054990.1 | - | 7 - 8 | 659 | 63 |
AK023151.1 | - | 4 - 5 | 362 | 63 | |
AK292485.1 | - | 7 - 8 | 690 | 63 | |
AK307494.1 | - | 8 - 9 | 866 | 63 | |
AK309772.1 | - | 6 - 7 | 543 | 63 | |
BC034357.1 | - | 7 - 8 | 671 | 63 | |
BC065481.1 | - | 7 - 8 | 650 | 63 | |
BC113585.1 | - | 7 - 8 | 717 | 63 | |
BC113587.1 | - | 7 - 8 | 717 | 63 | |
BX647450.1 | - | 7 - 8 | 670 | 63 | |
BX647992.1 | - | 4 - 5 | 318 | 63 |