Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016230.3 | NP_057314.2 | 1 - 2 | 206 | 98 |
GenBank mRNA | AK313146.1 | - | 1 - 2 | 179 | 98 |
AL832113.1 | - | 1 - 2 | 171 | 98 | |
BC025380.1 | - | 1 - 2 | 173 | 98 | |
DA274558.1 | - | 1 - 2 | 211 | 98 |